Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128170.2 | NP_001121642.1 | 7 | 1310 | ||
NM_001128171.2 | NP_001121643.1 | 7 | 1435 | |||
NM_001276279.1 | NP_001263208.1 | 7 | 1432 | |||
NM_173369.3 | NP_775545.1 | 6 | 1301 | |||
GenBank mRNA | AK039054.1 | - | 8 | 1439 | ||
AK122389.1 | - | 7 | 1416 | |||
AK133479.1 | - | 6 | 1265 | |||
AK147204.1 | - | 7 | 1279 | |||
AK147426.1 | - | 7 | 1274 | |||
AK167424.1 | - | 7 | 1396 | |||
BC042438.1 | - | 6 | 1254 | 3983771 |