Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028841.3 | NP_083117.2 | 6, 7 | 808 | ||
GenBank mRNA | AI019499.1 | - | 3, 4 | 279 | ||
AK008761.1 | - | 6, 7 | 753 | |||
AK166272.1 | - | 6, 7 | 811 | |||
AK171691.1 | - | 6, 7 | 757 | |||
BC010346.1 | - | 6, 7 | 753 | 2811935 | ||
CD740983.1 | - | 3, 4 | 377 |