Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028841.3 | NP_083117.2 | 1 | 247 | ||
GenBank mRNA | AK008761.1 | - | 1 | 192 | ||
AK166272.1 | - | 1 | 250 | |||
AK171691.1 | - | 1 | 196 | |||
BC010346.1 | - | 1 | 192 | 2811935 | ||
BY734818.1 | - | 1 | 247 |