Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028854.3 | NP_083130.1 | 9 | 872 | ||
GenBank mRNA | AK007086.1 | - | 9 | 960 | ||
BC100468.1 | - | 9 | 938 | 30916687 | ||
BC132329.1 | - | 9 | 872 | 40130606 | ||
BC138332.1 | - | 9 | 872 | 8861352 | ||
BC145239.1 | - | 9 | 869 | 9053791 | ||
BC145240.1 | - | 9 | 921 |