Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028866.3 | NP_083142.2 | 18 | 3224 | ||
GenBank mRNA | AB086191.1 | - | 17 | 3003 | ||
AK031786.1 | - | 4 | 1475 | |||
BC010638.1 | - | 5 | 1551 | |||
BC019526.1 | - | 3 | 364 |