Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028868.3 | NP_083144.1 | 9 | 1330 | ||
GenBank mRNA | AK010337.1 | - | 9 | 1331 | ||
AK083655.1 | - | 9 | 1335 | |||
AK146688.1 | - | 9 | 1414 | |||
AY099096.1 | - | 9 | 1302 | |||
BC012261.1 | - | 5 | 873 | |||
BC030938.1 | - | 9 | 1162 | 3980339 |