Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277121.1 | NP_001264050.1 | 3 | 357 | ||
NM_001277122.1 | NP_001264051.1 | 2 | 191 | |||
NM_001277123.1 | NP_001264052.1 | 2 | 191 | |||
NM_028871.2 | NP_083147.1 | 3 | 360 | |||
GenBank mRNA | AF441128.1 | - | 2 | 200 | ||
AK011307.1 | - | 2 | 109 | |||
AK133840.1 | - | 2 | 117 | |||
AK141145.1 | - | 3 | 305 | |||
AK144682.1 | - | 3 | 310 | |||
AK144992.1 | - | 2 | 109 | |||
AK145849.1 | - | 2 | 135 | |||
AK147043.1 | - | 3 | 326 | |||
AK152039.1 | - | 3 | 332 | |||
AK181037.1 | - | 1 | 53 | |||
BC004679.1 | - | 2 | 108 | |||
BC026850.1 | - | 3 | 293 | |||
BC038051.1 | - | 3 | 337 | 3500728 | ||
BY311081.1 | - | 2 | 166 |