Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277121.1 | NP_001264050.1 | 9 | 1280 | ||
NM_001277122.1 | NP_001264051.1 | 8 | 1119 | |||
NM_001277123.1 | NP_001264052.1 | 8 | 1114 | |||
NM_028871.2 | NP_083147.1 | 9 | 1283 | |||
GenBank mRNA | AF441128.1 | - | 8 | 1123 | ||
AK011307.1 | - | 8 | 1032 | |||
AK046495.1 | - | 2 | 148 | |||
AK133840.1 | - | 8 | 1040 | |||
AK144682.1 | - | 9 | 1233 | |||
AK144992.1 | - | 8 | 1037 | |||
AK145849.1 | - | 8 | 1058 | |||
AK147043.1 | - | 9 | 1250 | |||
AK152039.1 | - | 9 | 1255 | |||
BC004679.1 | - | 8 | 1031 | |||
BC026850.1 | - | 9 | 1214 | |||
BC038051.1 | - | 9 | 1260 | 3500728 |