Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277121.1 | NP_001264050.1 | 2 | 179 | ||
NM_028871.2 | NP_083147.1 | 2 | 182 | |||
GenBank mRNA | AF441128.1 | - | 1 | 22 | ||
AK141145.1 | - | 2 | 127 | |||
AK144682.1 | - | 2 | 132 | |||
AK147043.1 | - | 2 | 148 | |||
AK152039.1 | - | 2 | 154 | |||
BC026850.1 | - | 2 | 115 | |||
BC038051.1 | - | 2 | 159 | 3500728 |