Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028873.4 | NP_083149.3 | 2 | 212 | ||
GenBank mRNA | AK017830.1 | - | 2 | 226 | ||
AK032681.1 | - | 2 | 193 | |||
AK045345.1 | - | 2 | 269 | |||
AK045445.1 | - | 2 | 306 | |||
AK053314.1 | - | 2 | 274 | |||
AK082579.1 | - | 2 | 213 | |||
AK083878.1 | - | 2 | 223 | |||
AK159388.1 | - | 2 | 225 | |||
AK168935.1 | - | 2 | 303 | |||
BC011146.1 | - | 2 | 277 | 4016209 |