Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028878.3 | NP_083154.1 | 7, 8 | 1086 | ||
GenBank mRNA | AJ633679.1 | - | 7, 8 | 1095 | ||
AK014544.1 | - | 7, 8 | 1087 | |||
AK028784.1 | - | 7, 8 | 1087 | |||
AK128923.1 | - | 6, 7 | 1429 | |||
AK162399.1 | - | 6, 7 | 831 | |||
BC107411.1 | - | 7, 8 | 1087 | 40059042 | ||
BC107412.2 | - | 7, 8 | 1091 | 40059050 |