Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028887.3 | NP_083163.3 | 39 | 5134 | ||
GenBank mRNA | AK016419.1 | - | 14 | 1611 | ||
AK030247.1 | - | 7 | 716 | |||
AK129181.1 | - | 17 | 2131 | |||
BC044905.1 | - | 3 | 281 | |||
BC058205.1 | - | 5 | 594 | |||
BC058618.1 | - | 15 | 1786 | |||
BC062946.1 | - | 25 | 2979 | 30357469 | ||
BC076627.1 | - | 32 | 4097 |