Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026665.4 | NP_080941.3 | 10 | 1429 | ||
GenBank mRNA | AK014873.1 | - | 10 | 1376 | ||
AK014982.1 | - | 8 | 887 | |||
AK016484.1 | - | 10 | 1213 | |||
AK019414.1 | - | 10 | 1309 | |||
AK028408.1 | - | 9 | 1201 | |||
AK028458.1 | - | 10 | 1257 | |||
AK167523.1 | - | 9 | 1078 | |||
AY225093.1 | - | 10 | 1222 | |||
AY251192.1 | - | 10 | 1374 | |||
BC025588.1 | - | 9 | 1209 | |||
BC050785.1 | - | 8 | 940 | 6703205 | ||
BC056968.1 | - | 10 | 1673 |