Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165983.1 | NP_001159455.1 | 5 | 463 | ||
NM_001165984.1 | NP_001159456.1 | 5 | 438 | |||
NM_001165985.1 | NP_001159457.1 | 5 | 463 | |||
NM_001165986.1 | NP_001159458.1 | 5 | 438 | |||
NM_001165987.1 | NP_001159459.1 | 5 | 463 | |||
NM_001165988.1 | NP_001159460.1 | 5 | 438 | |||
NM_028475.2 | NP_082751.1 | 5 | 463 | |||
NM_153489.2 | NP_705693.2 | 5 | 463 | |||
GenBank mRNA | AF276967.1 | - | 5 | 398 | ||
AK014274.1 | - | 5 | 400 | |||
AK030218.1 | - | 5 | 404 | |||
AK031934.1 | - | 5 | 443 | |||
AK048286.1 | - | 5 | 464 | |||
AK079895.1 | - | 5 | 399 | |||
AK088702.1 | - | 5 | 461 | |||
AK152135.1 | - | 5 | 417 | |||
AK190891.1 | - | 3 | 257 | |||
AK208584.1 | - | 3 | 256 | |||
BC023906.1 | - | 5 | 454 | 5321969 | ||
BC029075.1 | - | 5 | 453 | 4527901 | ||
BC050910.1 | - | 4 | 385 | 6417527 |