Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165983.1 | NP_001159455.1 | 11 | 976 | ||
NM_001165984.1 | NP_001159456.1 | 11 | 936 | |||
NM_001165985.1 | NP_001159457.1 | 11 | 994 | |||
NM_001165986.1 | NP_001159458.1 | 11 | 969 | |||
NM_001165987.1 | NP_001159459.1 | 10 | 901 | |||
NM_001165988.1 | NP_001159460.1 | 10 | 876 | |||
NM_028475.2 | NP_082751.1 | 11 | 961 | |||
NM_153489.2 | NP_705693.2 | 10 | 901 | |||
GenBank mRNA | AF276967.1 | - | 10 | 836 | ||
AK016538.1 | - | 4 | 228 | |||
AK030218.1 | - | 11 | 902 | |||
AK031934.1 | - | 10 | 881 | |||
AK048286.1 | - | 11 | 977 | |||
AK051093.1 | - | 1 | 779 | |||
AK079895.1 | - | 11 | 930 | |||
AK082804.1 | - | 2 | 80 | |||
AK088702.1 | - | 11 | 992 | |||
BC023906.1 | - | 10 | 892 | 5321969 | ||
BC029075.1 | - | 11 | 951 | 4527901 | ||
BC050910.1 | - | 10 | 883 | 6417527 |