Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165983.1 | NP_001159455.1 | 15 | 1838 | ||
NM_001165984.1 | NP_001159456.1 | 15 | 1798 | |||
NM_001165985.1 | NP_001159457.1 | 15 | 1856 | |||
NM_001165986.1 | NP_001159458.1 | 15 | 1831 | |||
NM_001165987.1 | NP_001159459.1 | 14 | 1763 | |||
NM_001165988.1 | NP_001159460.1 | 14 | 1738 | |||
NM_028475.2 | NP_082751.1 | 15 | 1823 | |||
NM_153489.2 | NP_705693.2 | 14 | 1763 | |||
GenBank mRNA | AF276967.1 | - | 14 | 1698 | ||
AK016538.1 | - | 8 | 1090 | |||
AK030218.1 | - | 15 | 1764 | |||
AK031934.1 | - | 14 | 1743 | |||
AK048286.1 | - | 15 | 1839 | |||
AK051093.1 | - | 5 | 1642 | |||
AK079895.1 | - | 15 | 1795 | |||
AK082804.1 | - | 6 | 942 | |||
AK088702.1 | - | 15 | 1854 | |||
AK122210.1 | - | 2 | 201 | |||
AK208727.1 | - | 1 | 154 | |||
AK218166.1 | - | 1 | 154 | |||
BC023906.1 | - | 14 | 1754 | 5321969 | ||
BC029075.1 | - | 15 | 1813 | 4527901 | ||
BC050910.1 | - | 14 | 1745 | 6417527 |