Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028932.4 | NP_083208.1 | 1, 2 | 286 | ||
GenBank mRNA | AK016628.1 | - | 967 | |||
AK036661.1 | - | 1, 2 | 258 | |||
AK044181.1 | - | 1, 2 | 287 | |||
AK044737.1 | - | 1, 2 | 223 | |||
BC079658.1 | - | 1, 2 | 263 | 6853920 |