Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153501.2 | NP_705721.2 | 7 | 1380 | ||
GenBank mRNA | AK016755.1 | - | 9 | 1347 | ||
AK154023.1 | - | 5 | 738 | |||
AK154080.1 | - | 7 | 1344 | |||
AK159175.1 | - | 7 | 1372 | |||
AK167861.1 | - | 7 | 1088 | |||
AK213864.1 | - | 2 | 82 | |||
BC106184.1 | - | 7 | 1334 | 30241166 |