Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037221.2 | NP_001032298.1 | 3 | 1206 | ||
NM_001163433.1 | NP_001156905.1 | 2 | 686 | |||
NM_028966.3 | NP_083242.1 | 3 | 1206 | |||
GenBank mRNA | AK034323.1 | - | 2 | 800 | ||
AK133470.1 | - | 3 | 1185 | |||
AK134243.1 | - | 3 | 1204 | |||
AK161277.1 | - | 2 | 680 | |||
AK163751.1 | - | 2 | 1227 | |||
BC145827.1 | - | 3 | 1072 | 40131130 | ||
BC145829.1 | - | 3 | 1071 | 40131132 | ||
DQ278487.1 | - | 3 | 983 |