Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163635.1 | NP_001157107.1 | 25, 26 | 3585 | ||
GenBank mRNA | AK149368.1 | - | 11, 12 | 1550 | ||
BC063101.1 | - | 8, 9 | 1005 |