Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163635.1 | NP_001157107.1 | 12 | 1603 | ||
GenBank mRNA | AK047094.1 | - | 12 | 1462 |