Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159288.1 | NP_001152760.1 | 20 | 2619 | ||
NM_198162.1 | NP_937805.1 | 19 | 1946 | |||
GenBank mRNA | AK173042.1 | - | 20 | 2619 | ||
BC059243.1 | - | 19 | 1946 | 6417551 | ||
BC063082.1 | - | 8 | 886 |