Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028994.2 | NP_083270.1 | 5, 6 | 1061 | ||
GenBank mRNA | AK007429.1 | - | 2, 3 | 255 | ||
AK030501.1 | - | 5, 6 | 1062 | |||
AK031612.1 | - | 5, 6 | 906 | |||
AK034927.1 | - | 5, 6 | 940 | |||
AK147972.1 | - | 5, 6 | 915 | |||
AK165094.1 | - | 5, 6 | 939 | |||
AK170242.1 | - | 5, 6 | 915 | |||
BC010318.1 | - | 5, 6 | 924 | 3596227 | ||
BC023437.1 | - | 3, 4 | 529 |