Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028995.3 | NP_083271.1 | 3 | 473 | ||
GenBank mRNA | AK018640.1 | - | 3 | 450 | ||
AK049696.1 | - | 3 | 508 | |||
AK053937.1 | - | 476 | ||||
AK082376.1 | - | 475 | ||||
AK087811.1 | - | 3 | 476 | |||
AK138843.1 | - | 3 | 474 | |||
AK148206.1 | - | 3 | 492 |