Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyroxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029011.2 | NP_083287.2 | 5 | 745 | ||
GenBank mRNA | AK014668.1 | - | 5 | 475 | ||
AK154222.1 | - | 5 | 507 | |||
AK160239.1 | - | 5 | 461 |