Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029020.2 | NP_083296.2 | 1 | 355 | ||
GenBank mRNA | AK014319.1 | - | 1 | 86 | ||
AK014688.1 | - | 1 | 108 | |||
AK030624.1 | - | 1 | 82 | |||
AK038760.1 | - | 1 | 82 | |||
AK049152.1 | - | 1 | 99 | |||
AK076315.1 | - | 1 | 106 | |||
AK166704.1 | - | 1 | 89 | |||
AK170334.1 | - | 1 | 89 | |||
BC015301.1 | - | 1 | 355 | 4237894 |