Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029022.1 | NP_083298.1 | 4, 5 | 608 | ||
GenBank mRNA | AK014701.1 | - | 4, 5 | 602 | ||
AK028703.1 | - | 4, 5 | 582 | |||
AK143725.1 | - | 4, 5 | 605 | |||
AK165936.1 | - | 4, 5 | 605 | |||
BC015296.1 | - | 3, 4 | 542 | |||
BC139390.1 | - | 4, 5 | 603 | 8862412 | ||
BC139391.1 | - | 4, 5 | 603 | 8862413 |