Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029023.3 | NP_083299.3 | 12 | 1342 | ||
GenBank mRNA | AF330052.1 | - | 12 | 1298 | ||
AK014680.1 | - | 12 | 1343 | |||
AK050181.1 | - | 12 | 1280 | |||
AK153225.1 | - | 12 | 1330 | |||
AK153485.1 | - | 11 | 1131 | |||
BC004847.1 | - | 12 | 1332 | 2650587 | ||
BC021399.1 | - | 12 | 1297 | 4193674 |