Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029023.3 | NP_083299.3 | 10 | 987 | ||
GenBank mRNA | AF330052.1 | - | 10 | 943 | ||
AK014680.1 | - | 10 | 988 | |||
AK050181.1 | - | 10 | 925 | |||
AK153225.1 | - | 10 | 975 | |||
AK153485.1 | - | 9 | 776 | |||
BC004847.1 | - | 10 | 977 | 2650587 | ||
BC021399.1 | - | 10 | 942 | 4193674 |