Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109688.2 | NP_001103158.1 | 25 | 3200 | ||
NM_001286578.1 | NP_001273507.1 | 25 | 3106 | |||
NM_029081.3 | NP_083357.2 | 26 | 3278 | |||
NM_029897.2 | NP_084173.1 | 25 | 3227 | |||
GenBank mRNA | AK017577.1 | - | 8 | 852 | ||
AK029825.1 | - | 26 | 3198 | |||
AK162114.1 | - | 22 | 2780 | |||
AK163233.1 | - | 22 | 2623 | |||
AK172966.1 | - | 25 | 2960 | |||
BC027763.1 | - | 26 | 3054 | 5354483 | ||
BC028836.1 | - | 7 | 799 | |||
BC038611.1 | - | 2 | 629 | |||
BC049905.1 | - | 13 | 1444 | |||
BC053913.1 | - | 25 | 3054 | 30053530 |