Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhcr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053272.2 | NP_444502.2 | 2 | 375 | ||
GenBank mRNA | AK017937.1 | - | 2 | 137 | ||
AK083737.1 | - | 2 | 330 | |||
AK129036.1 | - | 2 | 375 | |||
AY039762.1 | - | 2 | 263 | |||
BC019797.1 | - | 2 | 319 | 5054108 |