Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205157.1 | NP_001192086.1 | 9 | 1052 | ||
NM_001205158.1 | NP_001192087.1 | 9 | 1042 | |||
NM_138303.2 | NP_612176.1 | 8 | 1190 | |||
GenBank mRNA | AK004959.1 | - | 8 | 976 | ||
AK033705.1 | - | 9 | 1001 | |||
AK133631.1 | - | 9 | 1012 | |||
BC002282.1 | - | 8 | 1105 | 3497525 | ||
BC003289.1 | - | 9 | 983 | 3583087 |