Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt8d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029102.3 | NP_083378.2 | 9, 10 | 1077 | ||
GenBank mRNA | AK003894.1 | - | 9, 10 | 1078 | ||
AK136351.1 | - | 9, 10 | 1059 | |||
BC082561.1 | - | 9, 10 | 1006 | 30603848 |