Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133858.4 | NP_598619.2 | 9 | 2228 | ||
NM_199475.1 | NP_955769.1 | 9 | 2255 | |||
GenBank mRNA | AB075603.1 | - | 8 | 1266 | ||
AK036838.1 | - | 9 | 2220 | |||
AK087859.1 | - | 9 | 2071 | |||
AK169673.1 | - | 9 | 2255 | |||
AK177378.1 | - | 1 | 39 | |||
BC008112.1 | - | 2 | 169 | |||
BC039762.1 | - | 8 | 1073 | |||
BC051048.1 | - | 8 | 1138 | |||
BC057901.1 | - | 7 | 757 | |||
BC069953.1 | - | 2 | 259 | |||
BC093521.1 | - | 5 | 824 | 4007022 | ||
BC094388.1 | - | 9 | 2072 | 5708548 | ||
BC132301.1 | - | 8 | 1500 | 40130578 | ||
BC144749.1 | - | 8 | 1500 | 9053297 |