Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133803.2 | NP_598564.2 | 18 | 2209 | ||
GenBank mRNA | AK031020.1 | - | 18 | 2174 | ||
AK087994.1 | - | 18 | 2173 | |||
AK150101.1 | - | 18 | 2169 | |||
AK150126.1 | - | 18 | 2163 | |||
AK166842.1 | - | 18 | 2174 | |||
AK168195.1 | - | 18 | 2162 | |||
AK218694.1 | - | 1 | 30 | |||
BC004600.1 | - | 17 | 2151 | 3584924 | ||
BC066817.1 | - | 18 | 2160 | 30429618 |