Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133803.2 | NP_598564.2 | 17 | 2081 | ||
GenBank mRNA | AK031020.1 | - | 17 | 2046 | ||
AK039975.1 | - | 13 | 3513 | |||
AK087994.1 | - | 17 | 2045 | |||
AK150101.1 | - | 17 | 2041 | |||
AK150126.1 | - | 17 | 2035 | |||
AK166842.1 | - | 17 | 2046 | |||
AK168195.1 | - | 17 | 2034 | |||
AK185970.1 | - | 1 | 142 | |||
AK200466.1 | - | 1 | 110 | |||
BC004600.1 | - | 16 | 2023 | 3584924 | ||
BC066817.1 | - | 17 | 2032 | 30429618 |