Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fggy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113412.1 | NP_001106883.1 | 2 | 227 | ||
NM_029347.2 | NP_083623.1 | 2 | 276 | |||
GenBank mRNA | AK085534.1 | - | 2 | 237 | ||
AK138439.1 | - | 2 | 224 | |||
BC031708.1 | - | 2 | 194 | 4194716 | ||
CB953569.1 | - | 2 | 277 |