Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178392.4 | NP_848479.1 | 3, 4 | 519 | ||
GenBank mRNA | AK012317.1 | - | 2, 3 | 176 | ||
AK138261.1 | - | 3, 4 | 520 | |||
AK152833.1 | - | 3, 4 | 525 | |||
BC030456.1 | - | 3, 4 | 501 | 5401615 |