Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rai14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166408.1 | NP_001159880.1 | 15 | 1436 | ||
NM_030690.3 | NP_109615.1 | 15 | 1522 | |||
GenBank mRNA | AF202315.1 | - | 14 | 1309 | ||
AF274866.1 | - | 15 | 1501 | |||
AK129333.1 | - | 15 | 1372 | |||
AK141233.1 | - | 11 | 1022 | |||
AK160119.1 | - | 14 | 1345 | |||
BC052458.1 | - | 15 | 1522 | 6830677 | ||
BU708854.1 | - | 5 | 532 |