Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081309.1 | NP_001074778.1 | 6 | 2226 | ||
GenBank mRNA | AK042953.1 | - | 5 | 2943 | ||
AK050333.1 | - | 6 | 2196 | |||
AK087938.1 | - | 6 | 2217 | |||
AK169508.1 | - | 5 | 3013 | |||
BC017537.1 | - | 5 | 1828 | |||
BC024083.1 | - | 3 | 700 | |||
BC157947.1 | - | 6 | 2051 | 9007281 | ||
BC158047.1 | - | 6 | 2051 | 9088160 |