Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081309.1 | NP_001074778.1 | 3, 4 | 1307 | ||
GenBank mRNA | AK042953.1 | - | 3, 4 | 1295 | ||
AK050333.1 | - | 3, 4 | 1277 | |||
AK054559.1 | - | 3, 4 | 1324 | |||
AK087938.1 | - | 3, 4 | 1298 | |||
AK169508.1 | - | 2, 3 | 2094 | |||
BC017537.1 | - | 2, 3 | 909 | |||
BC157947.1 | - | 3, 4 | 1132 | 9007281 | ||
BC158047.1 | - | 3, 4 | 1132 | 9088160 |