Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029397.3 | NP_083673.3 | 3 | 337 | ||
NM_170598.2 | NP_733486.2 | 3 | 305 | |||
GenBank mRNA | AF345334.1 | - | 2 | 93 | ||
AF393215.1 | - | 3 | 300 | |||
AF393216.1 | - | 3 | 268 | |||
AK035014.1 | - | 3 | 295 | |||
AK080772.1 | - | 3 | 295 | |||
BC052473.1 | - | 3 | 280 | 6833669 |
Gene Symbol: Cpne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC058662.1 | - | 3 | 241 |