Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081287.2 | NP_001074756.2 | 9, 10 | 921 | ||
NM_001161620.1 | NP_001155092.1 | 9, 10 | 921 | |||
GenBank mRNA | AK012883.1 | - | 9, 10 | 895 | ||
AK017344.1 | - | 12, 13 | 1283 | |||
AK033081.1 | - | 9, 10 | 881 | |||
AK036956.1 | - | 9, 10 | 892 | |||
AK078849.1 | - | 9, 10 | 920 | |||
BC049662.1 | - | 9, 10 | 875 | |||
BC117550.1 | - | 8, 9 | 768 | 40105182 | ||
BC118058.1 | - | 8, 9 | 768 | 40105180 |