Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030261.4 | NP_084537.2 | 4 | 676 | ||
GenBank mRNA | AK017464.1 | - | 4 | 573 | ||
AK155398.1 | - | 4 | 677 | |||
BC132086.1 | - | 4 | 497 | 40130363 | ||
BC138690.1 | - | 4 | 497 | 8861712 | ||
KF543243.1 | - | 4 | 425 |