Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027448.2 | NP_081724.1 | 3 | 809 | ||
NM_029434.3 | NP_083710.2 | 3 | 705 | |||
GenBank mRNA | AK015260.1 | - | 3 | 677 | ||
AK047956.1 | - | 3 | 672 | |||
BC049101.1 | - | 3 | 737 | 5702365 | ||
BC052060.1 | - | 3 | 737 | 5702365 | ||
CK639693.1 | - | 2 | 546 |