Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159640.2 | NP_001153112.1 | 12 | 1159 | ||
NM_001159641.2 | NP_001153113.1 | 11 | 1048 | |||
NM_175225.5 | NP_780434.1 | 12 | 1426 | |||
GenBank mRNA | AK015624.1 | - | 12 | 1146 | ||
AK036512.1 | - | 12 | 1382 | |||
AK132709.1 | - | 11 | 1063 | |||
AK146320.1 | - | 11 | 1031 | |||
AK160983.1 | - | 11 | 1294 | |||
BC024597.1 | - | 12 | 1146 | 4036044 |