Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029453.2 | NP_083729.2 | 24 | 2944 | ||
GenBank mRNA | AK015153.1 | - | 12 | 1177 | ||
AK015753.1 | - | 13 | 1397 |