Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290811.1 | NP_001277740.1 | 4 | 533 | ||
NM_029475.2 | NP_083751.1 | 4 | 425 | |||
GenBank mRNA | AK016299.1 | - | 4 | 530 | ||
AK049697.1 | - | 4 | 526 | |||
BC050879.1 | - | 4 | 377 | 5687835 | ||
BC052048.1 | - | 4 | 377 | 5687835 |