Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290811.1 | NP_001277740.1 | 6, 7 | 807 | ||
NM_001290812.1 | NP_001277741.1 | 4, 5 | 593 | |||
NM_029475.2 | NP_083751.1 | 6, 7 | 699 | |||
GenBank mRNA | AK016299.1 | - | 6, 7 | 804 | ||
AK039677.1 | - | 4, 5 | 583 | |||
AK049697.1 | - | 6, 7 | 800 | |||
BC050879.1 | - | 6, 7 | 651 | 5687835 | ||
BC052048.1 | - | 6, 7 | 651 | 5687835 |