Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029492.4 | NP_083768.4 | 1, 2 | 193 | ||
GenBank mRNA | AK017158.1 | - | 1, 2 | 194 | ||
AK031184.1 | - | 1, 2 | 152 | |||
AK049270.1 | - | 1, 2 | 173 | |||
AK139216.1 | - | 1, 2 | 152 | |||
AK140182.1 | - | 1, 2 | 154 | |||
AK151840.1 | - | 1, 2 | 208 | |||
AK163174.1 | - | 1, 2 | 191 | |||
AK170017.1 | - | 1, 2 | 211 | |||
AK170139.1 | - | 1, 2 | 134 | |||
AY668951.1 | - | 1, 2 | 111 | |||
BC019536.1 | - | 1, 2 | 210 | 4039734 |